ContactUsLinkComponent
Acid Neutralization Trap, Welch®
Catalog # 54994-092
Supplier:  Welch Vacuum
Product available on GSA Advantage®
undefined
Acid Neutralization Trap, Welch®
Catalog # 54994-092
Supplier:  Welch Vacuum
Supplier Number:  1420H-20
Product available on GSA Advantage®
Specifications
  • Description:
    Acid Neutralization Trap, 8×13"
  • Cat. no.:
    54994-092
  • Supplier No.:
    1420H-20
Specifications
About this item
The Acid Neutralization Trap contains a large alkaline element cartridge to neutralize hydrous acids before they enter the vacuum pump
  • ISO NW flange connection
  • Calcium Hydroxide element
  • Configure with prior cold trap to reduce loading
The element changes from white to bluish-transparent when spent and can be observed through the transparent trap body.