Order Entry
Canada
ContactUsLinkComponent
ULTROL® Grade, Tris Base Buffer, Calbiochem®, MilliporeSigma
ULTROL® Grade, Tris Base Buffer, Calbiochem®, MilliporeSigma
Catalog # CA80058-772
Supplier:  MilliporeSigma
CAS Number:  
ULTROL® Grade, Tris Base Buffer, Calbiochem®, MilliporeSigma
Catalog # CA80058-772
Supplier:  MilliporeSigma
Supplier Number:  648311-5KG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Fibre carton
  • Description:
    Tris base
  • Size:
    5 kg
  • Storage temperature:
    15...30 °C
  • Cat. no.:
    CA80058-772
  • pH:
    7.0 - 9.0
  • Supplier No.:
    648311-5KG

Specifications

About this item

Tris Base buffer is useful in the pH range of 10.3 to 11.3 and has a pKa of 8.1 at 25 °C.

Has an absorbance <0.05 (1.0 M, H₂O, 260 nm).