AlphaTec® 23-201 Chemical resistant gloves, Ansell
Catalog #: 75795-558
Supplier: ANSELL HEALTHCARE PRODUCTS LLC MX
Specifications
- Ambidextrous/Hand-specific:Hand-specific
- Glove Color:Blue
- Length:70 cm (27 1/2")
- Latex/Latex-free:Latex-free
- Thickness:4 mm
- Finish:Sandblast - Smooth
- Cat. no.:75795-558
- Supplier No.:124614
Specifications
About this item
Gloves offer dexterity and pliability at low temperatures, and provide good grip in handling both wet and slippery materials owing to the roughened sandblast finish.
- Seamless liner offers very good comfort in use
- Manufactured under EC quality assurance system
Ideal for use in automotive, food processing, and oil and gas industries for adjusting parts and systems, deboning, defeathering and skinning, preparation of site - unloading of lines and valves, primary cutting, steaming, frying, and roasting.
Certifications: Complies with the standards EN420 (General Requirements), EN388 (Mechanical Hazards) and EN374 (Chemical and Micro-organisms Hazards).
Packaging: Gloves are packaged in 12 pairs in a bag; 72 pairs in a carton.
Customers Who Bought This Also Bought