Shaker with 180° Shaking Motion, Glas-Col
Catalog # 33725-050
Supplier: Glas-Col
Small Business Enterprise
Specifications
- Shaking motion:180 degree shaking motion
- Shipping weight:11 lbs.
- Speed:2-30 rpm
- Cat. no.:33725-050
- Electrical:120 V
- Supplier No.:099AWS18012
Specifications
About this item
Simulates hand shaking by rotating through 180° arc.
- Shakes two flasks simutaneously
- Bench top mounting
- Use with screw cap or side arm flasks
For use with small to medium peptide synthesis flasks and culture flasks (up to 2-¹/₄" diameter, 10" long). This shaker offers 180 degree agitation shaking for solid phase peptide synthesis and for hard to dissolve solids such as polymers.
Ordering information: Shaker base with 2 large finger clamps.
Delivery: 7 to 10 days after order.