Holder for Bench Top Reciprocating Shaker, Mojonnier Fat Extraction Flask, Glas-Col
57020-299
: Glas-Col
Small Business Enterprise
- Capacity:25 ml
- Description:Flask Holder Rack for Mojonnier Fat Extraction Flask
- Environmentally Preferable:
- Cat. no.:57020-299
- Supplier No.:099AMFG3
This flask holder lets you use our bench top shaker for Mojonnier fat extraction.
- Two mojonnier flask holders can be shaken at once
- Load maximum of up to 5 pounds need not be balanced
- Use for vertical and horizontal shaking
Each holder accepts up to four flasks 37 mm in diameter. Unstoppered flasks may be shaken vertically. For horizontal shaking the flasks should be stoppered and held in with the built in retrainers.
: One holder holds four flasks.
: 7-10 days after order.