96-Well C8 Single-Break LUMITRAC™ 600 Strip Plate, Greiner Bio-One
Catalog # 10803-076
Supplier: Greiner Bio-One
Specifications
- Color:White
- Description:LUMITRAC™ 600 (High Binding) Microplate
- Cat. no.:10803-076
- Well Shape:C-Bottom
- Strip Format:1×8 well
- Supplier No.:705092
Specifications
About this item
Break-apart strips with frame.
- PS Material
- High binding
C-bottom (flat with curved edge) strips. White wells.