Corrugated SpongeCote Anti-Fatigue Mat, Wearwell®
Supplier: Wearwell
Certificates
About this item
The tight corrugations on the surface provide extra traction underfoot.
- Corrugated surface offers enhanced traction
- Fatigue-fighting sponge
- Easy to clean
- Ideal for industrial environments
Corrugated SpongeCote is a classic solution for durability and productivity enhancement. Popular long before Diamond-Plate surfaces were available, the tight corrugations on the surface provide extra traction underfoot. The Flexlink PVC sponge base is a budget friendly, fatigue fighting solution. Ideally suited for use in dry environments.
Certifications: Meets ASTM D395-98/20%.
Customers Who Bought This Also Bought