Order Entry
Canada
ContactUsLinkComponent
 
Threaded NMR Tubes, with Caps and Septa, Kimble Chase
Catalog # KT897635-0800
undefined
Threaded NMR Tubes, with Caps and Septa, Kimble Chase
Catalog # KT897635-0800
Supplier Number:  897635-0800

Specifications

  • Ext.Ø:
    5 mm
  • Description:
    NMR Tube, 8-425 Thread
  • Length:
    190.5 mm (7.5")
  • Cat. no.:
    KT897635-0800
  • Frequency:
    360 MHz
  • Supplier No.:
    897635-0800

Specifications

About this item

Ideal for use with air-sensitive samples and reaction studies where compounds may be added via syringe through the septum cap.

  • For use in instruments with field strengths greater than 360 MHz
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements