ALA Disks™ Delta-Aminolevulinic Acid, Hardy Diagnostics
Catalog # 75846-306
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:ALA Disks™, Delta-Aminolevulinic Acid
- Size:1x50 disks
- Cat. no.:75846-306
- Supplier No.:Z7081
Specifications
About this item
ALA Disks™, delta-aminolevulinic acid, for the differentiation and speciation of Haemophilus spp., a porphyrin test.
- A useful aid for differentiating species of Haemophilus
- Spring-loaded for smooth and reliable dispensing
- Single metal disk dispenser
Each cartridge contains 50 disks that are spring-loaded for smooth and reliable dispensing from the single metal disk dispenser by BD BioSciences.
ALA Disks require the use of a handheld 366 nm ultra violet lamp.