ContactUsLinkComponent
Compact Stand for Overhead Stirrers, Caframo
  CA58960-202
 :  Caframo
undefined
Compact Stand for Overhead Stirrers, Caframo
  CA58960-202
 :  Caframo
 :  A210
  • Rod Length:
    610 mm
  • Width:
    301.3 mm (11⅞")
  • Description:
    Compact Stand for Overhead Stirrers BDC2002 and BDC2010
  • Length:
    240 mm (9 ⁷⁄₁₆ in)
  • Cat. no.:
    CA58960-202
  • Supplier No.:
    A210
The compact stand is ideal for a small work space or under a fume hood.
The stand base is composed of cast zinc-aluminum alloy coated with chemical resistant epoxy.
 : Includes the stand base and 24" x 5/8" dia., 304 stainless steel solid rod.