Order Entry
Canada
Orders LinkContactUsLinkComponent
BBL™ M-PA-C Agar, BD

BBL™ M-PA-C Agar, BD

M-PA-C Agar, 500 g
Catalog # CA90005-076
Supplier:   BD
Supplier #:  298153
BBL™ M-PA-C Agar, BD
Catalog # CA90005-076
Supplier:  BD
CAS Number:  
BBL™ M-PA-C Agar, BD
BBL™ M-PA-C Agar, BD

M-PA-C Agar, 500 g

CAS Number:  
Price
$627.00
Log in for availability
Each (500g)
Product Details & Documents

M-PA-C Agar is used for the selective recovery and enumeration of Pseudomonas aeruginosa from water.

  • Contains yeast extract, lysine, and carbohydrates
  • Sodium chloride maintains osmotic equilibrium
  • Salts provide essential ions
  • Phenol red is a pH indicator
  • Kanamycin inhibits growth of gram-positive organisms

Colonies on membrane filters are counted using a stereoscopic microscope at 10-15x magnification.

Documents

Specifications

Specifications

  • Description:
    M-PA-C Agar
  • Size:
    500 g
  • Cat. No.:
    CA90005-076

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.