ContactUsLinkComponent
Difco™ MacConkey Agar Base, BD Diagnostics
Difco™ MacConkey Agar Base, BD Diagnostics
Catalog # CA90004-036
Supplier:  BD
CAS Number:  
undefined
Difco™ MacConkey Agar Base, BD Diagnostics
Catalog # CA90004-036
Supplier:  BD
Supplier Number:  281810
CAS Number:  
Specifications
  • Description:
    MacConkey Agar Base
  • Size:
    500 g
  • Cat. no.:
    CA90004-036
  • Supplier No.:
    281810
Specifications
About this item
MacConkey Agar Base is a solid medium recommended for use in qualitative procedures for the cultivation of gram-negative bacilli.
  • Peptones provide nitrogenous nutrients and amino acids
  • Sodium chloride maintains osmotic equilibrium
  • Crystal violet and bile salts inhibit most gram-positive organisms
MacConkey Agar Base is used with added carbohydrate to differentiate enteric gram-negative bacilli based on fermentation reactions.