Corning® Gosselin™ Storage Bottles, Round, HDPE, Corning
Supplier: Corning
Certificates
About this item
HDPE, translucent with screw caps.
- Bottles are supplied capped aseptic or ionised
- Excellent shock resistance
- Wide neck, easy to fill
A range of round wide mouth bottles. Ideal for use in laboratory, production plant or in the field for storage of liquids, solids and powders.
Frequently Bought Together