BBL™ Nutrient Broth, BD Life Sciences
Nutrient Broth, 5 mL
Supplier: BD
Supplier #: 221669
Jump to:
Product Details & Documents
Nutrient Broth is a general-purpose medium for the cultivation of nonfastidious bacteria.
- Peptone and beef extract provide essential nutrients
- Very simple nutrient formula
- Effective for nonfastidious microorganism growth
This relatively simple formulation supports the growth of nonfastidious microorganisms due to its content of peptone and beef extract.
Documents
Specifications
Specifications
- Description:Nutrient Broth
- Size:5 mL