ContactUsLinkComponent
BBL™ Cooked Meat Medium with Glucose, Hemin & Vitamin K, BD
BBL™ Cooked Meat Medium with Glucose, Hemin & Vitamin K, BD
Catalog # CA90004-616
Supplier:  BD
CAS Number:  
BBL™ Cooked Meat Medium with Glucose, Hemin & Vitamin K, BD
Catalog # CA90004-616
Supplier:  BD
Supplier Number:  295982
CAS Number:  
Specifications
  • Description:
    Cooked Meat Medium with Glucose, Hemin & Vitamin K
  • Size:
    9 mL Tubes
  • Cat. no.:
    CA90004-616
  • Supplier No.:
    295982
Specifications
About this item
Cooked Meat Medium and the enriched medium are used for the cultivation of anaerobes, especially pathogenic clostridia.
  • Muscle protein provides amino acids and other nutrients
  • Muscle provides reducing substances to permit growth of strict anaerobes
  • Growth is indicated by turbidity and presence of gas bubbles
Cooked Meat Medium provides a favorable environment for the growth of anaerobes. The sulfhydryl groups, which exert the reducing effect, are more available in denatured protein; therefore, the meat particles are cooked for use in the medium.