Sampling Probes Assemblies for Autosamplers, PerkinElmer
97013-184
- Description:FIAS Standard Sampling Probe Assembly, 1.0 mm I-Ø
- Cat. no.:97013-184
- Supplier No.:B3001772
Sampling probe assemblies for AS-90/90A/90plus/91/93plus/S10 autosampler.
Corrosion-resistant probes are suitable for inorganic acids and most organic solvents, except NMP.
PTFE probes provide laminar flow and are the most resistant to chemical degradation.
: Corrosion-resistant sampling probe assemblies includes sampling Probe with FEP tube, screw fitting and capillary tubing assembly.