Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Vortexer

Vortexer, 3.7 mm, 1000 - 3000 rpm, variable
  470222-698
 :  HS120209
Vortexer
  470222-698
Vortexer
Vortexer

Vortexer, 3.7 mm, 1000 - 3000 rpm, variable

 
$584.90
   
Each Case of 2
 
 

Multi-Purpose, Maximum Convenience

  • Variable speed (1000-3000 rpm) presets
  • Stable elastomeric device feet provide vibration dampening and prevent movement during operation
  • Head can be removed for cleaning purposes
  • High impact ABS body shell

This sleek and modern designed vortex mixer is capable of meeting the mixing requirements of the most popular test tubes used in today's labs without the need to swap out mixing head units. The innovative design makes maximum use or the space available, providing holding slots for no less that six different sizes of tube and a standard assay plate, while the central pad area can be used for quick hand-held mixing of a vast range of tubes and vessels. 110-120V, 50-60 Hz, 50 Watts, US/NA.

 : Two-year warranty

 
  • Description:
    Vortexer
  • Speed range:
    1000 - 3000 rpm, variable
  • Orbit:
    3.7 mm
  • Cat. No.:
    470222-698
  • Control:
    3 position slider —(top) intermittent, (middle) off, (bottom) continuous
  • Capacity:
    50 mL conical tubes x 2 (horizontally mounted);
    15 mL conical tubes x 2 (horizontally mounted);
    5 mL tubes x 2 (horizontally mounted in the same retention space as 15 mL);
    1.5/2 mL microfuge tubes x 4 (vertically mounted);
    0.5 mL microfuge tubes x 6 (vertically mounted);
    0.2 mL microfuge tubes (strip tubes) x 24 (vertically mounted)
  • W×D×H:
    171×190×197 mm
  • Weight:
    3.93 kg (8.66 lbs.)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.