Detachable Thermocouples, Process Reactors, Type T and K, Chemglass
Supplier: Chemglass
Certificates
About this item
The detachable thermocouples are for direct insertion in the reactors using a CG-1048-A or CG-1042-E thermocouple adapter, while the 4” long Hastelloy C-276 style is for use with the CG-1971 baffles and adapters.
- Thermocouples for use with Chemglass process reactors
- Available in both “T” (blue connections) and “K” (yellow connections)
Detachable type thermocouples have a standard male plug (OST) and require a coiled extension cord for connection to the CG-3498 temperature monitor
Note: The 1/2” dia. probes have a transition to 6” of PTFE leads and are clear FEP PTFE coated.
The 4” long Hastelloy C-276 thermocouples are supplied with a 4ft FEP insulated cord having a standard male plug (OST).
Customers Who Bought This Also Bought