Order Entry
Canada
ContactUsLinkComponent
PhosphoSafe™ Extraction Reagent, MilliporeSigma
PhosphoSafe™ Extraction Reagent, MilliporeSigma
Catalog # CA82601-054
Supplier:  MilliporeSigma
CAS Number:  
PhosphoSafe™ Extraction Reagent, MilliporeSigma
Catalog # CA82601-054
Supplier:  MilliporeSigma
Supplier Number:  71296-4
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Description:
    PhosphoSafe™ extraction buffer
  • Size:
    125 ml
  • Storage temperature:
    ≤–70 °C
  • Cat. no.:
    CA82601-054
  • Application:
    Extracts cytosolic proteins from mammalian and insect cells
  • Supplier No.:
    71296-4

Specifications

About this item

PhosphoSafe™ extraction buffer efficiently extracts cytosolic proteins from mammalian and insect cells while preserving their phosphorylation state.

This reagent is compatible with kinase assays, protein interaction analysis, and other applications. This reagent is provided in convenient 25 ml bottles.

Delivery: This reagent includes four phosphatase inhibitors: sodium fluoride, sodium vanadate, β-glycerophosphate, and sodium pyrophosphate.