You searched for: Dialysis Tubing
Its ability to selectively diffuse molecules across a semi-permeable membrane gives dialysis tubing its important place in classic laboratory technique. Dialysis is used for a wide variety of applications: desalting, buffer exchange, removal of labeling reagents, drug-binding studies, cell growth and feeding, virus purification, and blood treatment. Created from a natural source of cellulose, dialysis tubing provides a reliable source for separating molecules based on size for testing purposes, and no pre-treatment is required before use.
Dialysis Tubing
Supplier: Ward's Science
The semi-permeable membrane, created from a natural source of cellulose, allows passage of particles up to 14000 Da.
Expand 15 Items
Pierce SnakeSkin™ Pleated Dialysis Tubing, MWCO 3500 to 10000, Thermo Scientific
Supplier: Thermo Scientific
Simplifies dialysis of sample volumes between 15 ml and 100 ml.
Expand 7 Items
Pierce™ Slide-A-Lyzer™ Dialysis Flasks, Thermo Scientific
Supplier: Thermo Scientific
Pierce™ Slide-A-Lyzer™ Dialysis Flasks facilitate simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules in sample volumes up to 250 ml.
Expand 4 Items
Spectra/Por® Biotech Regenerated Cellulose Dialysis Tubing, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Biotech RC is crafted from a regeneration process that yields superior temperature tolerances and chemical compatibilities with the same high purity and selectivity as Biotech CE.
Expand 6 Items
Spectra/Por® 4, Dialysis Membranes, MWCO 12000 to 14000, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes.
Expand 6 Items
Spectra/Por® Biotech Cellulose Ester Dialysis Membranes, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Biologically inert and ultra-pure, Biotech Cellulose Ester (CE) Membrane offers the largest range of concise MWCO's (100 to 1000000 Daltons) for desalting, isolating ionic species and macromolecular purifications.
Expand 27 Items
Spectra/Por® Micro Float-A-Lyzer Dialysis Devices, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Spectrum introduces the new Micro Float-A-Lyzer® to expand our popular line of ready-to-use dialysis devices for the easy and efficient dialysis of very small sample volumes (100 to 500 µl).
Expand 14 Items
Spectra/Por® 5, Dialysis Tubing, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated.
Expand 2 Items
Spectra/Por® 7 Dialysis Membranes, MWCO 1000 to 50000
Supplier: Spectrum Laboratories
Free of trace metal and sulfur for separations involving a possible sensitivity to such materials.
Expand 41 Items
Biotech Cellulose Ester Dialysis Membrane Tubing Trial Kits
Supplier: Spectrum Laboratories
Ideal for small volume dialysis or evaluation of membrane for an application, biotech grade Cellulose Ester (CE) tubing is now available in 1 m length trial kits with a pair of tubing closures.
Expand 32 Items
Spectra/Por® 3 Dialysis Membranes, MWCO 3500, Spectrum® Laboratories
Supplier: Spectrum Laboratories
For dialysis and ultrafiltration in biochemical work.
Expand 4 Items
Spectra/Por® 1-5 Standard RC Dry Dialysis Trial Kits, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Ideal for small volume dialysis or evaluation of membrane for an application, multi-purpose standard Regenerated Cellulose (RC) tubing is now available in 1 and 5 m length trial kits with a pair of tubing closures.
Expand 19 Items
Spectra/Por® 1 Dialysis Membranes, MWCO 6000 to 8000, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Standard Regenerated Cellulose (RC) is composed of natural cellulose regenerated from cotton linters, therefore these membranes are perfectly suited for a broad range of laboratory dialysis.
Expand 8 Items
Spectra/Por® in vivo Micro-Dialysis Hollow Fibers, Spectrum® Laboratories
Supplier: Spectrum Laboratories
The implantation of individual micro-dialysis hollow fibers is an ideal step in the recovery or introduction of micro-quantity biologicals or pharmaceutical materials in a localized tissue environment.
Expand 1 Items
Spectra/Por Regenerated Cellulose Dialysis Discs and Sheets, Spectrum Laboratories
Supplier: Spectrum Laboratories
Pre-cut single layer membrane discs and sheets for a variety of dialysis equipment.
Expand 9 Items
Pierce™ Slide-A-Lyzer™ G2 Dialysis Cassettes, Thermo Scientific
Supplier: Thermo Scientific
The Slide-A-Lyzer® G2 Dialysis Cassette offers efficiency, convenience and sample protection in one package. Sample loading and removal are easily accomplished by using a serological pipette or hypodermic needle attached to a syringe. The built-in air chamber provides sample buoyancy and vertical orientation of the cassette during dialysis.
Expand 26 Items
Spectra/Por® Dialysis Tubing, Regenerated Cellulose, MWCO 12000 to 14000, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Standard dialysis tubing for all laboratory dialysis applications.
Expand 3 Items
Dialysis Tubing Funnels
Supplier: Bel-Art
Reduce difficulty in handling dialysis bags with these convenient devices.
Expand 1 Items
Spectra/Por®, Dialysis Tubing, Spectrum® Laboratories
Supplier: Spectrum Laboratories
The dialysis tubing is sealed at one end with a closure and open at the other.
Expand 2 Items
Spectra/Por® 6 Dialysis Membranes, MWCO 1000 to 50000, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Available in wet form in 0.05% sodium azide as a preservative only.
Expand 51 Items
Spectra/Por® Float-A-Lyzer G2 Dialysis Devices, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Spectrum introduces the Spectra/Por® Float-A-Lyzer® G2, the next generation in Ready-to-Use laboratory dialysis devices featuring proprietary Ultra-pure Biotech Cellulose Ester (CE) Membrane.
Expand 27 Items
Pierce™ Slide-A-Lyzer® Dialysis Cassettes, Thermo Scientific
Supplier: Thermo Scientific
For sample dialysis, low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and concentration. This type of membrane is compatible with a number of common chemicals and buffers. The colour-coded transparent frames of the Slide-A-Lyzer® cassette allow instant recognition of the MWCO of the membrane and make viewing the needle easier during sample injection.
Expand 34 Items
Spectra/Por® 2 Dialysis Membranes, MWCO 12000 to 14000, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Standard Regenerated Cellulose (RC) is composed of natural cellulose regenerated from cotton linters, therefore these membranes are perfectly suited for a broad range of laboratory dialysis.
Expand 5 Items
Pierce™ Slide-A-Lyzer® MINI Dialysis Units, Thermo Scientific
Supplier: Thermo Scientific
Versatile, disposable units are ideal for sample preparation to dialyze proteins or nucleic acids.