

2-Propanol ≥99.85%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Boiling Pt:77…83 °C (1.013 hPa)
Melting Pt:–90 °C
Density:0.784…0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
Specifications
Specification Test Results
- IR spectrum:complying
- Assay (GC):Min. 99.85 %
- Non-volatile matter:Max. 0.0003 %
- Water (Karl Fischer):Max. 0.05 %
- Free acid (as C₂H₅COOH):Max. 0.001 %
- Absorbance at 400 nm:Max. 0.01
- Absorbance at 260 nm:Max. 0.01
- Absorbance at 230 nm:Max. 0.10
- Absorbance at 220 nm:Max. 0.20
- Absorbance at 210 nm:Max. 0.70
- Absorbance at 205 nm:Max. 1.0
- APHA:Max. 10
- Vapor Pressure:44 mmHg ( 25 °C)
- Refractive Index:n20/D 1.377 (lit.)
Specification Test Results
Related Information
Frequently Bought Together