Orders LinkContactUsLinkComponent
Twin Bucket, Contec®
Twin Bucket, Contec®
Twin divided bucket with lid, stainless steel wringer and casters, Green, 20 L (5.2 gal.)
Catalog # 76270-914
Supplier:   CONTEC, INC. MX
Supplier #:  2743S
Twin Bucket, Contec®
Catalog # 76270-914
Supplier:  CONTEC, INC. MX
Twin Bucket, Contec®
Twin Bucket, Contec®
Twin divided bucket with lid, stainless steel wringer and casters, Green, 20 L (5.2 gal.)
Price
$589.10
New Customer Promotional Price
Log in for availability
Case of 1
Product Details & Documents
The Twin Bucket is a divided bucket with a curved stainless steel wringer.
  • Ideal for smaller controlled environments and labs with limited storage space
  • Designed to work with the curved stainless steel wringer
  • Fully autoclavable (casters are non-autoclavable)
The Twin Bucket is made of durable polypropylene, with a well -fitting lid to eliminate liquid spillage during transport. Use one side of the bucket for cleaning and disinfecting solutions, and the other for rinsing.
Delivery information: Includes 4 casters for added mobility.
Specifications
  • Capacity:
    20 L (5.2 gal.)
  • Color:
    Green
  • Description:
    Twin divided bucket with lid, stainless steel wringer and casters