QualakoteESD® PDWS Wave Solder Gloves, ESD-Safe, QRP
: PIP
The QRP PDWS Qualakote ESD Low Heat Wave Solder Gloves are extra thick for thermal protection.
- Nylon + conductive carbon/nylon yarn
- Nitrile foamed palm dip for extra toughness
- Seamless knit, open back
- Human Body Model ANSI/ESD SP15.1 CAFE` resistance: 10⁷ uncoated area; 10⁸¯⁹ coated area
- Machine washable for economy of repeated use
PDWS Qualakote ESD Wave Solder Glove is knit from nylon carbon yarn and features a Nitrile anti-static dip palm for dependable grip. Heat protection, yet thin for maximum dexterity. Seamless knit for greater comfort. Surface resistivity 10⁶ ohms.
- Catalog No:
- 89041-082
- 89041-080
- 89041-076
- 89041-078
- 89041-074
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Finish:
- Nitrile foamed palm dip
- Nitrile foamed palm dip
- Nitrile foamed palm dip
- Nitrile foamed palm dip
- Nitrile foamed palm dip
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Anti-static:
- +
- +
- +
- +
- +
Frequently Bought Together