PolyTuff® 25G PolyTuff Static Dissipative Gloves, QRP
Supplier: PIP
Certificates
About this item
The unique, form-flexible PolyTuff polyurethane (1.5 mil) film wicks perspiration away for greater comfort.
- Packaged for ISO 5 (Class 100) applications
- Anti-static 10⁹ ohms per square surface unit
- Resists common solvents such as MeCl, TCE, and IPA
- ESD safe for Class II devices
- Ergonomic Comfort Curve™ hand specific design
RoHS Compliant, 1.5 mil film. Static dissipative, non-generative ISO 5 (Class 100) glove; dissipates induced charges in under 0.25 seconds. Seamless Comfort Curve paired RH/LH gloves minimize hand and finger fatigue. Resistant to many common solvents, including TCE and IPA. MIL-B-81705 complaint. ISO 5 (Class 100) processed and packaged in double-bagged, static safe packaging. Suggested for Class I and certain Class II static sensitive devices.
Specifications
- Catalog No:
- 89041-054
- 32891-350
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Powdered/Powder-free:
- Powder-free
- Powder-free