PolyTuff® 20G PolyTuff Solvent Gloves, QRP
Supplier: PIP
Certificates
About this item
Excellent resistance to punctures, snags, and abrasion, coupled with high tactile sensitivity.
- Packaged for ISO 5 (Class 100) applications
- Protection from chlorinated solvents, (MeCL and TCE), acetone, xylene, freons, and IPA
- Tough, chemical & solvent resistant - solvent process polyurethane, sulfur-free
- Ergonomic Comfort Curve™ hand specific design
Ultra-tough, 5 mil, 12" length.
Applications: Precision parts cleaning, precision assembly requiring chemical resistance, solvents, etching; industrial paint and finish applications, automotive, aircraft maintenance, degreasing operations, general cleaning; electro-chemical polishing, optics, disk drives.
Permeation tested per ASTM F-739. Seamless, anatomically accurate comfort curve design minimizes finger fatigue and delivers enhanced flexibility, hand specific. ISO 5 (Class 100) processed and packaged.
Specifications
- Catalog No:
- 32891-132
- 89041-052
- 32891-065
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free