Order Entry
Canada
ContactUsLinkComponent
 
CoolBigPrep™ Adapter, MP Biomedicals, LLC
Catalog # CA76046-182
Supplier:  MP Biomedicals
undefined
CoolBigPrep™ Adapter, MP Biomedicals, LLC
Catalog # CA76046-182
Supplier:  MP Biomedicals
Supplier Number:  116002531
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Capacity:
    2 Tubes
  • Description:
    CoolBigPrep™ 2×50 ml Adapter
  • Size:
    2×50 ml
  • Sample throughput:
    Low, Medium
  • Adapter compatibility:
    FastPrep®-24 Adapters, Cryogenic, Large Volume
  • Instrument Compatibility:
    FastPrep-24; FastPrep-96 (requires ConeFlex™ Legacy Adapter)
  • Kit compatibility:
    FastDNA 50 ml Spin Kit for Soil; and all maxiprep 50 ml
  • Tube compatibility:
    50 ml
  • Key applications:
    Extractions of nucleic acids, proteins, metabolites and other small molecules from virtually any temperature unstable or sensitive sample. Larger sized Sample Holder
  • Material of construction:
    Composite
  • Storage and handling:
    Store at Room Temperature (15...30 °C)
  • Sample volume:
    up to 40 g
  • Sample type:
    Soil; Plants; Animal Tissues; Feces; Yeast; Bacteria (Gram +/-); Fungi; Spores; Wastewater; Food; Seeds; Roots
  • Weight:
    700 g
  • Product type:
    Instruments & Systems
  • Cat. no.:
    CA76046-182

Specifications

About this item

CoolBigPrep™ 2×50 ml Adapter is ideally suited for extractions of any temperature unstable or sensitive biological compounds such as RNA, siRNA, metabolites, intermediates, and enzymes from even the hardest samples to lyse.

  • Consistent, reproducible results
  • For use with FastPrep-24 or FastPrep-96 Homogenizer
  • Ensure efficient cooling of samples

CoolBigPrep™ 2×50 ml Adapter is a cryogenic adapter which allows simultaneous lysis of 2×50 ml samples

Use with FastPrep-24 or FastPrep-96 (with ConeFlex Legacy Adapter), MP Bio Lysing Matrix Tubes and/or FastPrep purification kits to lyse, grind or homogenize samples under cryogenic conditions, for the extraction and purification of nucliec acids, proteins, metabolites and other small molecules and more.

Based on passive temperature control technology, the CoolBigPrep™ adapter ensures an efficient cooling of the samples as dry-ice, placed into the moving tray is in direct contact with the sample tubes. Due to a high heat transfer capacity and FastPrep® precise settings of lysis parameters, the samples can be repeatably homogenized in their unthawed state. A wide range of disposable 50 ml Lysing Matrix tubes ensure thorough homogenization of any sample type in 40 seconds or less.