ContactUsLinkComponent
Costar® Transtar-96® Liquid Transfer System, Corning
Costar® Transtar-96® Liquid Transfer System, Corning
Catalog # 29442-530
Supplier:  Corning
Product available on GSA Advantage®
undefined
Costar® Transtar-96® Liquid Transfer System, Corning
Catalog # 29442-530
Supplier:  Corning
Supplier Number:  7605
Product available on GSA Advantage®
Specifications
  • Volume:
    25 - 200 µl
  • Description:
    96 Well Liquid Transfer System
  • Cat. no.:
    29442-530
  • Accuracy:
    ±5%
  • Supplier No.:
    7605
Specifications
About this item
The Costar® Transtar-96® System is a portable, autoclavable liquid handling system designed for use with 96 well plates.
  • Ideal for changing cell culture media and screening monoclonal antibodies
  • Enables liquids to be aspirated, transferred and dispensed