Order Entry
Canada
Orders LinkContactUsLinkComponent
Plastic Well Slides

Plastic Well Slides

Plastic well slides, dia. 2-3 mm, depth 16 mm, 10 slides, 3×1"
Catalog # 470218-416
Supplier #:  CSPLG1
Plastic Well Slides
Catalog # 470218-416
Plastic Well Slides
Plastic Well Slides

Plastic well slides, dia. 2-3 mm, depth 16 mm, 10 slides, 3×1"

Price
$11.05
Log in for availability
Pack of 10
Product Details & Documents

Reusable slides for liquid or aquatic specimens.

  • Quality slide making supplies
  • Hands-on scientific learning

Great for viewing your favorite algae, protist, or small aquatic invertebrate organisms.

Packaging: These reusable slides are sold as pack of 10 individual slides in a bag.

Specifications
  • Description:
    Plastic well slides, dia. 2-3 mm, depth 16 mm, 10 slides
  • L×W:
    3×1"