ContactUsLinkComponent
ISO-BB Column Test Mix, Restek
  75835-392
 :  Restek
 :  
ISO-BB Column Test Mix, Restek
  75835-392
 :  Restek
 :  35290
 :  
  • Concentration:
    250 µg/ml each in methylene chloride
  • Description:
    ISO-BB Column Test Mix
  • Size:
    1 ml/Ampule
  • Cat. no.:
    75835-392
  • Supplier No.:
    35290
8 components