Order Entry
Canada
ContactUsLinkComponent
 
Volatile Amine Column Test Mix, Restek
Catalog # 75835-334
Supplier:  Restek
CAS Number:  
Volatile Amine Column Test Mix, Restek
Catalog # 75835-334
Supplier:  Restek
Supplier Number:  35008
CAS Number:  

Specifications

  • Concentration:
    In methanol:dichloromethane (50:50)
  • Description:
    Volatile Amine Column Test Mix
  • Size:
    1 ml/Ampule
  • Cat. no.:
    75835-334
  • Supplier No.:
    35008

Specifications

About this item

8 components