Order Entry
Canada
ContactUsLinkComponent
XIL-350 Column Test Mix, Restek
XIL-350 Column Test Mix, Restek
Catalog # 75835-362
Supplier:  Restek
CAS Number:  
XIL-350 Column Test Mix, Restek
Catalog # 75835-362
Supplier:  Restek
Supplier Number:  35226
CAS Number:  

Specifications

  • Concentration:
    350 µg/ml each in methylene chloride
  • Description:
    XIL-350 Column Test Mix
  • Size:
    1 ml/Ampule
  • Cat. no.:
    75835-362
  • Supplier No.:
    35226

Specifications

About this item

8 components