LC OQ Wavelength Accuracy Standard , Restek
75834-604
: Restek
:
- Concentration:10 mg/ml in water 5 ml/ampule
- CAS No.:14017-55-1
- Description:erbium perchlorate
- Size:5 ml/Ampule
- Formula:Cl₃ErO₁₂
- Description:LC OQ Wavelength Accuracy Standard
- Cat. no.:75834-604
- Supplier No.:31053