Glycerin Standard Mix (3 components), Restek
75834-860
: Restek
:
- Concentration:In P and T methanol
- Description:Glycerin Standard Mix
- Size:1 ml/Ampule
- Cat. no.:75834-860
- Supplier No.:31891
Testing of glycerin for diethylene glycol with diethylene glycol (DEG) and ethylene glycol limit standards.
This guidance emphasizes the importance of screening raw material for the presence of diethylene glycol. Under cGMPs, drug manufacturers, not just glycerin manufacturers, must test glycerin prior to use to prevent DEG contamination in finished products. The FDA has worked extensively with the USP to modify the glycerin monograph, and these standards support the revised USP method.
Consists of diethylene glycol (0.5 mg/ml), ethylene glycol 0.5 and glycerin 20.
: Meet FDA guidance for industry.