European Pharmacopoeia/ICH Class 2 Mix B, Revised (10 Components), Restek
Catalog # 75835-452
Supplier: Restek
CAS Number:
Specifications
- Concentration:Prepared in water:dimethyl sulfoxide (80:20)
- Description:European Pharmacopoeia/ICH Class 2 Mix B, Revised
- Size:1 ml/Ampule
- Cat. no.:75835-452
- Supplier No.:36270
Specifications
About this item
The European pharmacopoeia/ICH Class 2 mix B, revised consists of 10 components, they are acetonitrile (410 μg/ml), chloroform (60 μg/ml), 1,2-dimethoxyethane (100 μg/ml), N,N-dimethylacetamide (1,090 μg/ml), 1,4-dioxane (380 μg/ml), 2-hexanone (50 μg/ml), methanol (3,000 μg/ml), nitromethane (50 μg/ml), pyridine (200 μg/ml) and tetralin (100 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.