Order Entry
Canada
ContactUsLinkComponent
Residual Solvents Class 2 - Mix B (8 Components), Restek
Residual Solvents Class 2 - Mix B (8 Components), Restek
Catalog # 75835-464
Supplier:  Restek
CAS Number:  
Residual Solvents Class 2 - Mix B (8 Components), Restek
Catalog # 75835-464
Supplier:  Restek
Supplier Number:  36280
CAS Number:  

Specifications

  • Concentration:
    In dimethyl sulfoxide
  • Description:
    Residual Solvents Class 2 - Mix B
  • Size:
    1 ml/Ampule
  • Cat. no.:
    75835-464
  • Supplier No.:
    36280

Specifications

About this item

The solvents consists of (C6) n-hexane (290 μg/ml), chloroform (60 μg/ml), 1,2-dimethoxyethane (100 μg/ml), 2-hexanone (50 μg/ml), nitromethane (50 μg/ml), pyridine (200 μg/ml), tetralin (100 μg/ml), trichloroethene (80 μg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.