Order Entry
Canada
ContactUsLinkComponent
SOM01.1 Deuterated Monitoring Compound Mix SIM Compounds (2 Components), Restek
SOM01.1 Deuterated Monitoring Compound Mix SIM Compounds (2 Components), Restek
Catalog # 75835-234
Supplier:  Restek
CAS Number:  
SOM01.1 Deuterated Monitoring Compound Mix SIM Compounds (2 Components), Restek
Catalog # 75835-234
Supplier:  Restek
Supplier Number:  33913
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in methylene chloride
  • Description:
    SOM01.1 Deuterated Monitoring Compound Mix SIM Compounds
  • Size:
    1 ml/Ampule
  • Cat. no.:
    75835-234
  • Supplier No.:
    33913

Specifications

About this item

The deuterated monitoring compound mix consists of fluoranthene-d10 and 2-methylnaphthalene-d10.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.