Minnesota Ag List 1 Pesticides Mix A (16 components), Restek
Catalog # 75834-950
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 ppm each in acetone
- Description:Minnesota Ag List 1 Pesticides Mix A
- Size:1 ml/Ampule
- Cat. no.:75834-950
- Supplier No.:32406
Specifications
About this item
Contains: Acetochlor, alachlor, atrazine, cyanazine (bladex), desethylatrazine, desisopropylatrazine, dimethenamid, ethalfluralin, metolachlor, metribuzin, pendimethalin, prometon, propachlor, propazine, simazine, trifluralin.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Ordering information: *Added to Minnesota Department of Agriculture List 1 pesticide (neutrals) incident investigation requirements, effective January 1, 2000.¹ CAS # 87674-68-8 manufactured by several companies under various trade names.
¹Analytical Lists for Pesticide Incident Investigations, Minnesota Department of Agriculture, Guidance Document 26 (3/99), St. Paul, MN. For a copy, visit their website at: www.mda.state.mn.us
¹Analytical Lists for Pesticide Incident Investigations, Minnesota Department of Agriculture, Guidance Document 26 (3/99), St. Paul, MN. For a copy, visit their website at: www.mda.state.mn.us