Test mix E1618 (13 components), Restek
Catalog # 75835-710
Supplier: Restek
CAS Number:
Specifications
- Concentration:0.05% volume/volume each in methylene chloride
- Description:E1618 Test Mix
- Size:1 ml/Ampule
- Cat. no.:75835-710
- Supplier No.:31613
Specifications
About this item
Components in this mix (0.5 μl/ml or 0.05% volume/volume each) are at 10X the concentration of the final test solution specified in ASTM 1618 and ASTM 1387.
The mix consists of (C6) n-hexane, (C8) n-octane, (C10) n-decane, (C12) n-dodecane, (C14) n-tetradecane, (C16) n-hexadecane, (C18) n-octadecane, (C20) n-eicosane, 2-ethyltoluene, 3-ethyltoluene, toluene, 1,2,4-trimethylbenzene, p-xylene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.