Pesticide Standard Mix B (11 Components), Restek
Catalog # 75834-946
Supplier: Restek
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:In hexane:toluene (90:10)
- Description:Pesticide Standard Mix B
- Size:1 ml/Ampule
- Cat. no.:75834-946
- Supplier No.:32298
Specifications
About this item
Consists of 11 components, those are aldrin, β-bhc, δ-bhc, cis-chlordane, trans-chlordane, 4,4'-dde, endosulfan ii, endosulfan sulfate, endrin aldehyde, endrin ketone and heptachlor epoxide.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.