Canadian Drinking Water OP Pesticides Mix (9 Components), Restek
Catalog # 75834-818
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml each
- Matrix:Acetonitrile
- Description:Canadian Drinking Water OP Pesticides Mix
- Size:1 ml/Ampule
- Cat. no.:75834-818
- Supplier No.:31867
Specifications
About this item
Consists of nine components, those are azinphos methyl, chlorpyrifos, diazinon, dimethoate, malathion, parathion, phorate, temephos and terbufos .
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.