Order Entry
Canada
ContactUsLinkComponent
Method 525.2 PCB Congener Mix (8 Components), Restek
Method 525.2 PCB Congener Mix (8 Components), Restek
Catalog # 75834-968
Supplier:  Restek
CAS Number:  
Method 525.2 PCB Congener Mix (8 Components), Restek
Catalog # 75834-968
Supplier:  Restek
Supplier Number:  32420
CAS Number:  

Specifications

  • Concentration:
    200 µg/ml each
  • Matrix:
    Acetone
  • Description:
    Method 525.2 PCB Congener Mix
  • Size:
    1 ml/Ampule
  • Cat. no.:
    75834-968
  • Supplier No.:
    32420

Specifications

About this item

Contains eight components, those are 2-chlorobiphenyl, 2,3-dichlorobiphenyl, 2,4,5-trichlorobiphenyl, 2,2',4,4'-tetrachlorobiphenyl, 2,2',3',4,6-pentachlorobiphenyl, 2,2',4,4',5,6'-hexachlorobiphenyl, 2,2',3,3',4,4',6-heptachlorobiphenyl and 2,2',3,3',4,5',6,6'-octachlorobiphenyl.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.