Order Entry
Canada
ContactUsLinkComponent
ASTM D3606 Backflush Standard (2,2,4-Trimethylpentane (isooctane)), Restek
ASTM D3606 Backflush Standard (2,2,4-Trimethylpentane (isooctane)), Restek
Catalog # 75834-576
Supplier:  Restek
CAS Number:  
ASTM D3606 Backflush Standard (2,2,4-Trimethylpentane (isooctane)), Restek
Catalog # 75834-576
Supplier:  Restek
Supplier Number:  30671
CAS Number:  

Specifications

  • Concentration:
    5% vol/vol in nonane
  • Description:
    2,2,4-Trimethylpentane (isooctane)
  • Size:
    1 ml
  • Cat. no.:
    75834-576
  • Supplier No.:
    30671

Specifications

About this item

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.