Sulfur Simulated Distillation Standard (SSDS), Restek
Catalog # 75835-128
Supplier: Restek
CAS Number:
Specifications
- Description:Sulfur Simulated Distillation Standard (SSDS)
- Size:1 ml
- Cat. no.:75835-128
- Supplier No.:33049
Specifications
About this item
The standard consists of ethanethiol, 1-propanethiol, 1-butanethiol, 1-pentanethiol, 1-hexanethiol, 1-heptanethiol, 3,5-dimethylbenzenethiol, 1-octanethiol, 1-nonanethiol, 1-decanethiol, 1-pentadecanethiolol, 1-hexadecanethiol and 1-octadecanethiol.