TNRCC 1006 Retention Time Marker Mix (9 Components), Restek
75834-774
: Restek
:
- Concentration:200 µg/ml in pentane
- Pack type:Ampoule
- Description:TNRCC 1006 Retention Time Marker Mix
- Size:1 ml
- Cat. no.:75834-774
- Supplier No.:31814
This TNRCC 1006 retention Ttme marker mix contains: (C6) n-Hexane, (C7) n-Heptane, (C8) n-Octane, (C10) n-Decane, (C12) n-Dodecane, (C16) n-Hexadecane, (C21) n-Heneicosane, (C28) n-Octacosane, (C35) n-Pentatriacontane.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.