Order Entry
Canada
ContactUsLinkComponent
MA VPH Matrix Spike Mix with Surrogate Rev. 1.1 (July 2004) (16 Components), Restek
MA VPH Matrix Spike Mix with Surrogate Rev. 1.1 (July 2004) (16 Components), Restek
Catalog # 75835-594
Supplier:  Restek
CAS Number:  
MA VPH Matrix Spike Mix with Surrogate Rev. 1.1 (July 2004) (16 Components), Restek
Catalog # 75835-594
Supplier:  Restek
Supplier Number:  30605
CAS Number:  

Specifications

  • Concentration:
    50 µg/ml in P and T methanol
  • Pack type:
    Ampoule
  • Description:
    MA VPH Matrix Spike Mix With Surrogate Rev. 1.1 (July 2004)
  • Size:
    1 ml
  • Cat. no.:
    75835-594
  • Supplier No.:
    30605

Specifications

About this item

The mix consists of (C5) n-pentane, (C9) n-nonane, (C10) n-decane, benzene, n-butylcyclohexane, 2,5-dibromotoluene (SS), ethylbenzene, 2-methylpentane, methyl tert-butyl ether, naphthalene, toluene, 1,2,4-trimethylbenzene, 2,2,4-trimethylpentane, m-xylene, o-xylene, p-xylene.

  • For current Massachusetts VPH method - surrogate included
  • Matrix spike mix at 50 μg/ml, per updated method
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.