MA VPH Standard with Surrogate Rev. 1.1 (July 2004) (16 Components), Restek
Catalog # 75835-592
Supplier: Restek
CAS Number:
Specifications
- Concentration:10000 µg/ml in P and T methanol
- Pack type:Ampoule
- Description:MA VPH Standard With Surrogate Rev. 1.1 (July 2004)
- Size:1 ml
- Cat. no.:75835-592
- Supplier No.:30604
Specifications
About this item
The mix consists of (C5) n-pentane, (C9) n-nonane, (C10) n-decane, benzene, n-butylcyclohexane, 2,5-dibromotoluene, ethylbenzene, 2-methylpentane, methyl tert-butyl ether, naphthalene, toluene, 1,2,4-trimethylbenzene, 2,2,4-trimethylpentane, m-xylene, o-xylene, p-xylene.
- Standard for current Massachusetts VPH method - surrogate included
- More analyses per ampul-standard with surrogate is 10000 μg/ml
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.