MA VPH Matrix Spike Mix with Surrogate (14 Components), Restek
Catalog # 75835-562
Supplier: Restek
CAS Number:
Specifications
- Concentration:2500 µg/ml each in P and T methanol
- Pack type:Ampoule
- Description:MA VPH Matrix Spike Mix With Surrogate
- Size:1 ml
- Cat. no.:75835-562
- Supplier No.:30454
Specifications
About this item
The MA VPH matrix spike mix consists of 14 components they are (C5) n-pentane, (C9) n-nonane, benzene, 2,5-dibromotoluene (SS), ethylbenzene, 2-methylpentane, methyl tert-butyl ether (mtbe), naphthalene, toluene, 1,2,4-trimethylbenzene, 2,2,4-trimethylpentane (isooctane), m-xylene, o-xylene and p-xylene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.