Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Recombinant Caspase 2

Human Recombinant Caspase 2

25 units
Catalog # CA82022-058
Supplier:   G-Biosciences
Supplier #:  RCH-002
Human Recombinant Caspase 2
Catalog # CA82022-058
Supplier:  G-Biosciences
CAS Number:  
Human Recombinant Caspase 2

25 units

CAS Number:  
Price
$2,379.91
Log in for availability
Each (250ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

G-Biosciences' Recombinant Human Caspases 1-3 and 6-10 are supplied lyophilized in quantities of 25 units

G-Biosciences' Recombinant Human Caspases 1-3 and 6-10 are members of the ICE (interleukin-1 β converting enzyme) family of cysteine proteases. They exist in cells as inactive pro-enzymes. During apoptosis, pro-caspases 1-3 and 6-10 are processed at aspartate residues by self-proteolysis and/or cleavage by another caspase. The matured (processed) form of these caspases consists of large and small subunits, which associate to form a heterotetramer active enzyme.

G-Biosciences' active recombinant caspase-2 cleaves caspase-2 chromogenic substrate Ac-VDVAD-pNA, and the signal can be detected spectrophotometrically.

G-Biosciences' active caspase-6 and caspase-7 have been shown in the involvement of the proteolysis of poly (ADP-ribose) polymerase (PARP), an enzyme that is involved in DNA repair and genomic maintenance. The active recombinant caspase-6 cleaves the substrates consisting of consensus sequence VEID (e.g. VEID-AFC and VEID-pNA).

One unit of the specific recombinant caspase is the enzyme activity that cleaves one nanomole of a specific caspase substrate per hour at 37°C in a reaction solution containing 50mM HEPES (at a pH of 7.2), 50mM NaCl, 0.1% CHAPS, 10mM EDTA, 5% Glycerol, and 10mM DTT.

Ordering information: Accessories available

Specifications

Specifications

  • Size:
    25 units
  • Cat. No.:
    CA82022-058
  • Enzyme type:
    Recombinant
  • Species:
    Human
  • Enzyme name:
    Caspase 2

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.