ContactUsLinkComponent
TSX Series High-Performance Plasma Freezers, Automatic Defrost, Thermo Scientific
undefined
TSX Series High-Performance Plasma Freezers, Automatic Defrost, Thermo Scientific
  • Catalog No:
  • 76320-248
  • 76320-318
  • 75839-098
  • 76320-246
  • 76208-606
  • 76208-608
  • 76320-320
  • Model:
  • TSX1230LD
  • TSX3030LA
  • TSX2330LA
  • TSX1230LA
  • TSX5030LA
  • TSX5030LD
  • TSX3030LD
  • Environmentally Preferable:
  • Energy Efficient
  • Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Energy Efficient
  • Cat. No.:
  • 76320-248
  • 76320-318
  • 75839-098
  • 76320-246
  • 76208-606
  • 76208-608
  • 76320-320
  • Electrical:
  • 208 - 230 V, 50/60 Hz, 15 A
  • 115 V, 60 Hz, 15 A
  • 115 V, 60 Hz, 15 A
  • 115 V, 60 Hz, 15 A
  • 115 V, 60 Hz, 20 A
  • 208 - 230 V, 50/60 Hz, 15 A
  • 208 - 230 V, 50/60 Hz, 15 A
  • Shelves/drawers:
  • 7 drawers
  • 7 drawers
  • 7 drawers
  • 7 drawers
  • 14 drawers
  • 14 drawers
  • 7 drawers
  • Door type:
  • Solid doors
  • Solid doors
  • Solid doors
  • Solid doors
  • Solid doors
  • Solid doors
  • Solid doors
  • Environmental certification:
  • Energy Star; EPA SNAP Compliant
  • Energy Star; EPA SNAP Compliant
  • Energy Star; EPA SNAP Compliant
  • Energy Star; EPA SNAP Compliant
  • Energy Star; EPA SNAP Compliant
  • Energy Star; EPA SNAP Compliant
  • Energy Star; EPA SNAP Compliant
Frequently Bought Together