SP Wilmad-LabGlass Stainless Steel Quick Release Clamps, SP Industries
Supplier: Avantor
Certificates
About this item
Made entirely from stainless steel, these clamps firmly hold reaction vessels with Duran® flanges and lids together
Quick release mechanism has a screw adjustment for tension control of flange.
Customers Who Bought This Also Bought